Order Entry
Czech Republic
ContactUsLinkComponent
Anti-HLA Class I Mouse Monoclonal Antibody [clone: W6/32] (HRP (Horseradish Peroxidase))
Anti-HLA Class I Mouse Monoclonal Antibody [clone: W6/32] (HRP (Horseradish Peroxidase))
Catalog # ABCAAB33256-50
Supplier:  Abcam
undefined
Anti-HLA Class I Mouse Monoclonal Antibody [clone: W6/32] (HRP (Horseradish Peroxidase))
Catalog # ABCAAB33256-50
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    HLA Class 1
  • Clonality:
    Monoclonal
  • Clone:
    W6/32
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a
  • Seq:
    1202
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pack type:
    Vial
  • Pk:
    50 µG

Specifications

About this item

HRP Mouse monoclonal [W6/32] to HLA Class I.

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: HLA Class I
Clonality: Monoclonal
Clone: W6/32
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Clone W6/32 recognises a conformational epitope, reacting with HLA class I alpha 3 and alpha 2 domains
Host: Mouse
Isotype: IgG2a
Reactivity: Human;Baoon;Cat;Bovine;Cynomolgus monkey;Macaque;Monkey