Anti-Semaphorin 7a Mouse Monoclonal Antibody [clone: MEM-150] (FITC (Fluorescein Isothiocyanate))
Catalog # ABCAAB26012-50
Supplier: Abcam
Specifications
- Antibody type:Primary
- Antigen name:Semaphorin 7a
- Antigen symbol:SEMA7a
- Clonality:Monoclonal
- Clone:MEM-150
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgM
- Reactivity:Human
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 tests
- Pack type:Vial
- Pk:50 Tests
Specifications
About this item
FITC Mouse monoclonal [MEM-150] to Semaphorin 7a.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Semaphorin 7a
Clonality: Monoclonal
Clone: MEM-150
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human