Anti-HIV1 Nef Mouse Monoclonal Antibody [clone: 3F2]
Catalog # ABCAAB25866-100
Supplier: Abcam
Specifications
- Antibody type:Primary
- Antigen symbol:HIV1 Nef
- Clonality:Monoclonal
- Clone:3F2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Western blot:Yes
- Seq:1286
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pack type:Vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [3F2] to HIV1 Nef.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: HIV1 Nef
Clonality: Monoclonal
Clone: 3F2
Conjugation:
Epitope: Mapped to amino acids 31 - 35 (GAASR, HIV - 1 HxB2)
Host: Mouse
Isotype: IgG1
Reactivity: HIV