Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-Tau 13 Mouse Monoclonal Antibody [clone: B11E8]
Katalogové číslo ABCAAB19030-50
Dodavatel:  Abcam
Anti-Tau 13 Mouse Monoclonal Antibody [clone: B11E8]
Katalogové číslo ABCAAB19030-50
Dodavatel:  Abcam

Specifikace

  • Antibody type:
    Primary
  • Antigen symbol:
    Tau 13
  • Clonality:
    Monoclonal
  • Clone:
    B11E8
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µl
  • Pk:
    50 µl

Specifikace

O tomto produktu

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Tau 13
Clonality: monoclonal
Clone: B11E8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human