Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Transferrin Receptor Mouse Monoclonal Antibody [clone: MEM-75] (PE (Phycpoerythrin))
Anti-Transferrin Receptor Mouse Monoclonal Antibody [clone: MEM-75] (PE (Phycpoerythrin))
Catalog # ABCAAB18242-50
Supplier:  Abcam
undefined
Anti-Transferrin Receptor Mouse Monoclonal Antibody [clone: MEM-75] (PE (Phycpoerythrin))
Catalog # ABCAAB18242-50
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    transferrin receptor
  • Antigen symbol:
    TFRC
  • Clonality:
    Monoclonal
  • Clone:
    MEM-75
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Seq:
    0780
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 Tests
  • Pack type:
    Vial
  • Pk:
    50 Tests

Specifications

About this item

PE Mouse monoclonal [MEM-75] to Transferrin Receptor.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Transferrin Receptor
Clonality: Monoclonal
Clone: MEM-75
Conjugation: PE (Phycpoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human