Order Entry
Czech Republic
ContactUsLinkComponent
Anti-BRCA1 Mouse Monoclonal Antibody [clone: MS13]
Anti-BRCA1 Mouse Monoclonal Antibody [clone: MS13]
Catalog # ABCAAB16781-100
Supplier:  Abcam
undefined
Anti-BRCA1 Mouse Monoclonal Antibody [clone: MS13]
Catalog # ABCAAB16781-100
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Breast cancer 1, early onset
  • Antigen symbol:
    BRCA1
  • Clonality:
    Monoclonal
  • Clone:
    MS13
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Seq:
    1286
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pack type:
    Vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [MS13] to BRCA1.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: BRCA1
Clonality: Monoclonal
Clone: MS13
Conjugation:
Epitope: Within the N-terminal 304 amino acids of BRCA1
Host: Mouse
Isotype: IgG1
Reactivity: Human