Anti-HIV1 p24 Mouse Monoclonal Antibody [clone: 39/5.4A]
Catalog # ABCAAB9071-100
Supplier: Abcam
Specifications
- Antibody type:Primary
- Antigen symbol:HIV1 p24
- Clonality:Monoclonal
- Clone:39/5.4A
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Caution:
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: HIV1 p24
Clonality: monoclonal
Clone: 39/5.4A
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: