Anti-Cytokeratin 10 Mouse Monoclonal Antibody [clone: DE-K10]
Catalog # ABCAAB9026-100
Supplier: Abcam
Specifications
- Antibody type:Primary
- Antigen name:keratin 10
- Antigen symbol:KRT10
- Clonality:Monoclonal
- Clone:DE-K10
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Western blot:Yes
- Seq:1286
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pack type:Vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [DE-K10] to Cytokeratin 10 - Cytoskeleton Marker.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Cytokeratin 10
Clonality: Monoclonal
Clone: DE-K10
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog;Human