Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Cyclophilin 40 Rabbit Polyclonal Antibody
Anti-Cyclophilin 40 Rabbit Polyclonal Antibody
  ABCAAB3562-100
 :  Abcam
undefined
Anti-Cyclophilin 40 Rabbit Polyclonal Antibody
  ABCAAB3562-100
 :  Abcam

 

  • Antibody type:
    Primary
  • Antigen name:
    peptidylprolyl isomerase D
  • Antigen symbol:
    PPID
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Gel shift:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Seq:
    1286
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

Rabbit polyclonal to Cyclophilin 40.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Cyclophilin 40
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat; Human