Specifications
- Antibody type:Primary
- Antigen name:Cholinergic receptor, muscarinic 2
- Antigen symbol:CHRM2
- Clonality:Monoclonal
- Clone:31-1D1
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Pig,Human,Rat,Mouse
- Western blot:Yes
- Seq:1310
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:125 µl
- Pack type:Vial
- Pk:125 µl
Specifications
About this item
Mouse monoclonal [31-1D1] to Muscarinic Acetylcholine Receptor 2/CM2.
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Muscarinic Acetylcholine Receptor 2
Clonality: monoclonal
Clone: 31-1D1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Pig, Rat