Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Adenovirus type 1 hexon E11 Mouse Monoclonal Antibody [clone: 1-M-13]
Anti-Adenovirus type 1 hexon E11 Mouse Monoclonal Antibody [clone: 1-M-13]
  ABCAAB2596-100
 :  Abcam
undefined
Anti-Adenovirus type 1 hexon E11 Mouse Monoclonal Antibody [clone: 1-M-13]
  ABCAAB2596-100
 :  Abcam

 

  • Antibody type:
    Primary
  • Antigen name:
    Adenovirus type 1 hexon E11
  • Clonality:
    Monoclonal
  • Clone:
    1-M-13
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Western blot:
    Yes
  • Seq:
    1286
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pack type:
    Vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [1-M-13] to Adenovirus type 1 hexon E11.

 : All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Adenovirus type 1 hexon E11
Clonality: Monoclonal
Clone: 1-M-13
Conjugation:
Epitope: The exact epitope recognised by this antibody has not been mapped
Host: Mouse
Isotype: IgG2a
Reactivity: Human adenovirus