Anti-Cyclin A2 Mouse Monoclonal Antibody [clone: E67.1]
Catalog # ABCAAB39-50
Supplier: Abcam
Specifications
- Antibody type:Primary
- Antigen name:cyclin A2
- Antigen symbol:CCNA2
- Clonality:Monoclonal
- Clone:E67.1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pack type:Vial
- Pk:50 µG
Specifications
About this item
Mouse monoclonal [E67.1] to Cyclin A2.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Cyclin A2
Clonality: Monoclonal
Clone: E67.1
Conjugation:
Epitope: The epitope for this antibody was mapped by phage selection - selected phage contained the sequence LDYP. This would place the epitope within the region spanning amino acids 144 - 148 of human Cyclin A2.Cyclin As that contain the epitopeCow A2Human A2Hamster A2Mouse A2 Cyclin As that do not contain the epitope Chicken A2Frog A2Human A1Mouse A1Frog A1.
Host: Mouse
Isotype: IgG2a
Reactivity: Human