Anti-IFN Alpha 2a Rabbit Polyclonal Antibody
Catalog # BIRBORB97718-20
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:IFN alpha 2a
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rabbit
- Western blot:Yes
- Form:lyophilized
- Storage buffer:Derived from a PBS containing solution
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:IgG Anti Human Interferon alpha 2a is developed in rabbits using Recombinant Human Interferon a 2a expressedin plants.
- Size:20 µg
- Pk:20 µG
Specifications
About this item
Western Blot: 1:500-1:1000
Type: Primary
Antigen: IFN Alpha 2a
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rabbit