Anti-CLIC4 Rabbit Polyclonal Antibody
Catalog # BIRBORB96056-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:chloride intracellular channel 4
- Antigen symbol:CLIC4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Storage buffer:PBS, pH 7,4, 150 mM sodium chloride, 0,02% sodium azide, 50% glycerol
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
ELISA: 1:40000; Immunochemistry: 10 ?g/ml; Western Blot: 1:500-1:1000
Type: Primary
Antigen: CLIC4 (chloride intracellular channel 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat