Anti-SLC6A3 Rabbit Polyclonal Antibody
Catalog # BIRBORB87686-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 6 (neurotransmitter transporter), member 3
- Antigen symbol:Solute carrier family 6, member 3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Storage buffer:PBS, 0,01% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
ELISA: 1 ?g/ml; Immunochemistry: 20 ?g/ml; Western Blot: 1 ?g/ml
Type: Primary
Antigen: SLC6A3 (solute carrier family 6 (neurotransmitter transporter), member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human