Anti-SLC7A2 Rabbit Polyclonal Antibody
Catalog # BIRBORB86967-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 7 (cationic amino acid transporter, y+ system), member 2
- Antigen symbol:Solute carrier family 7, member 2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Storage buffer:PBS, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
- ELISA: 1:1000; Immunochemistry: 10 μg/ml
Type: Primary
Antigen: SLC7A2 (solute carrier family 7 (cationic amino acid transporter, y+ system), member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human