Anti-SLC7A11 Rabbit Polyclonal Antibody
Catalog # BIRBORB86275-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 7 (anionic amino acid transporter light chain, xc- system), member 11
- Antigen symbol:Solute carrier family 7, member 11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Storage buffer:PBS, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Synthetic 19 amino acid peptide from near N-terminus of human SLC7A11
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
- Immunochemistry: 8 μg/ml
Type: Primary
Antigen: SLC7A11 (solute carrier family 7 (anionic amino acid transporter light chain, xc- system), member 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human