Anti-SLC7A5 Rabbit Polyclonal Antibody
BIRBORB85996-50
: Biorbyt
- Antibody type:Primary
- Antigen name:solute carrier family 7 (amino acid transporter light chain, L system), member 5
- Antigen symbol:Solute carrier family 7, member 5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Storage buffer:PBS, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Synthetic 15 amino acid peptide from internal region of human SLC7A5
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Immunochemistry: 15 ?g/ml
Type: Primary
Antigen: SLC7A5 (solute carrier family 7 (amino acid transporter light chain, L system), member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human