Anti-CISH Rabbit polyclonal antibody unconjugated
Katalogové číslo BIRBORB76299-100
Dodavatel: Biorbyt
Specifikace
- Antibody type:Primary
- Antigen name:cytokine inducible SH2-containing Protein
- Antigen symbol:CISH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Form:lyophilized
- Amino acid number:241 to 258
- Storage buffer:5mg BSA, 0,9mg NaCl, 0,2mg Na2HPO4, 0,05mg Thimerosal, 0,05mg NaN3.
- Sequence:LPLPRRMADYLRQYPFQL
- Storage temperature:Cool
- Concentration:500ug/ml,
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human CISH(241-258aa LPLPRRMADYLRQYPFQL), identical to the related rat and mouse sequences.
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifikace
O tomto produktu
0.1-0.5 ?g/ml
Type: Primary
Antigen: CISH (cytokine inducible SH2-containing protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: