Anti-CLCA1 Rabbit Polyclonal Antibody
Katalogové číslo BIRBORB69755-100
Dodavatel: Biorbyt
Specifikace
- Antibody type:Primary
- Antigen name:chloride channel accessory 1
- Antigen symbol:CLCA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifikace
O tomto produktu
Type: Primary
Antigen: CLCA1 (chloride channel accessory 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human