Order Entry
Czech Republic
ContactUsLinkComponent
Anti-SLC12A6 Rabbit Polyclonal Antibody
Anti-SLC12A6 Rabbit Polyclonal Antibody
Catalog # BIRBORB137942-100
Supplier:  Biorbyt
undefined
Anti-SLC12A6 Rabbit Polyclonal Antibody
Catalog # BIRBORB137942-100
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    solute carrier family 12 (potassium/chloride transporter), member 6
  • Antigen symbol:
    Solute carrier family 12, member 6
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    lyophilized
  • Storage buffer:
    5mg BSA, 0,9mg NaCl, 0,2mg Na2HPO4, 0,05mg Thimerosal, 0,05mg NaN3.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminal of human SLC12A6, different from the related mouse and rat sequences by one amino acid.
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: SLC12A6 (solute carrier family 12 (potassium/chloride transporter), member 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat