Order Entry
Czech Republic
ContactUsLinkComponent
Anti-NRG1 isoform-10 Rabbit Polyclonal Antibody
Anti-NRG1 isoform-10 Rabbit Polyclonal Antibody
Supplier:  Biorbyt
Certificates
undefined
Anti-NRG1 isoform-10 Rabbit Polyclonal Antibody
Supplier:  Biorbyt

Specifications

  • Catalog No:
  • BIRBORB128463-100
  • BIRBORB128463-200
  • Antigen symbol:
  • NRG1
  • NRG1
  • Antigen name:
  • NRG1 isoform-10
  • NRG1 isoform-10
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Purification:
  • purified by affinity chromatography
  • purified by affinity chromatography
  • Concentration:
  • 1 mg/ml
  • 1 mg/ml
  • Storage buffer:
  • PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
  • PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together