Anti-CLCNKA Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
Certifikáty
O tomto produktu
Type: Primary
Antigen: CLCNKA (chloride channel, voltage-sensitive Ka)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Specifikace
- Catalog No:
- BIRBORB125584-100
- BIRBORB125584-200
- Antigen symbol:
- CLCNKA
- CLCNKA
- Antigen name:
- chloride channel, voltage-sensitive Ka
- chloride channel, voltage-sensitive Ka
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes
- Purification:
- purified by affinity chromatography
- purified by affinity chromatography
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage buffer:
- PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifikace
Frequently Bought Together