Anti-SLC7A11 Rabbit Polyclonal Antibody (Cy5®)
Katalogové číslo BIRBORB122949-100
Supplier: Biorbyt
Specifications
- Druh protilátky:Primární
- Název antigenu:solute carrier family 7 (anionic amino acid transporter light chain, xc- system), member 11
- Symbol antigenu:Solute carrier family 7, member 11
- Klonovalita:Polyclonal
- Spojení:Cy5®
- Hostitel:Rabbit
- Izotyp:IgG
- Formulář:liquid
- Skladovací pufr:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molekulární hmotnost:52 kDa
- Skladovací teplota:Cool
- Transportní teplota:Ice
- Čištění:purified by affinity chromatography
- Rozměry protilátky:100 µl
- Bal.:100 µl
Specifications
About this item
Type: Primary
Antigen: SLC7A11 (solute carrier family 7 (anionic amino acid transporter light chain, xc- system), member 11)
Clonality: polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: