Anti-14-3-3 delta/zeta Rabbit Polyclonal Antibody
BIRBORB106463-100
: Biorbyt
- Antibody type:Primary
- Antigen symbol:14-3-3 delta/zeta
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Storage buffer:Phosphate buffer saline, pH. 7.4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Molecular weight:28 kDa
- Storage temperature:Cool
- Concentration:20 ug: 0,2 mg/ml; 50 ug: 0,5 mg/ml; 100 ug: 1 ug/ul; 200 ug: 1 ug/ul
- Shipping temperature:Ice
- Immunogen:A synthesized peptide derived from human 14-3-3 zeta/delta
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200; Immunofluorescence: 1:100-1:500
Type: Primary
Antigen: 14-3-3 delta/zeta (14-3-3 delta/zeta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat