Anti-HPV (Type 1, 6, 11, 16, 18, 31) Mouse Monoclonal Antibody
Catalog # BIRBORB27223-3
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:HPV (Type 1, 6, 11, 16, 18, 31)
- Antigen symbol:HPV 1/6/11/16/18/31
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG
- Storage temperature:Cool
- Concentration:neat
- Shipping temperature:Ice
- Immunogen:SDS disintegrated bovine Papillomavirus Type 1
- Size:3 ml
- Pk:3 mL
Specifications
About this item
Type: Primary
Antigen: HPV (Type 1, 6, 11, 16, 18, 31)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: