Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Mouse CD44 (Ly-24) Rat Monoclonal Antibody (FITC (Fluorescein))
Anti-Mouse CD44 (Ly-24) Rat Monoclonal Antibody (FITC (Fluorescein))
Catalog # BIRBORB26403-0.5
Supplier:  Biorbyt
Anti-Mouse CD44 (Ly-24) Rat Monoclonal Antibody (FITC (Fluorescein))
Catalog # BIRBORB26403-0.5
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    CD44
  • Clonality:
    Monoclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2b
  • Reactivity:
    Mouse
  • Storage buffer:
    0,01 M PBS, pH 7,4 containing 150 mM Sodium Chloride and 1% BSA.
  • Storage temperature:
    Cool
  • Concentration:
    0,5 mg/ml (OD280)
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    500 μg
  • Pk:
    0,5 mg

Specifications

About this item

Flow Cytometry: 1 ?g

Type: Primary
Antigen: Mouse CD44 (Ly-24)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG2b (Rat)
Reactivity: Mouse