Order Entry
Czech Republic
ContactUsLinkComponent
Anti-HIV-1 p55/17 Mouse monoclonal antibody [clone: 3A1]
Anti-HIV-1 p55/17 Mouse monoclonal antibody [clone: 3A1]
Catalog # BIRBORB24122-100
Supplier:  Biorbyt
Anti-HIV-1 p55/17 Mouse monoclonal antibody [clone: 3A1]
Catalog # BIRBORB24122-100
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    HIV1 p55/17
  • Clonality:
    Monoclonal
  • Clone:
    3A1
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Storage buffer:
    PBS, pH 7,2
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    Recombinant Protein
  • Purification:
    purified by ammonium sulfate precipitation
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Western Blot: 1:1000

Type: Primary
Antigen: HIV-1 p55/17
Clonality: monoclonal
Clone: 3A1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: