Anti-HPV 16 (E7) Mouse Monoclonal Antibody
Catalog # BIRBORB24091-1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Human Papilloma Virus
- Antigen symbol:HPV 16 (E7)
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a
- Western blot:Yes
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Concentration:4,1 mg/ml (OD280, E(0,1%)=1,4)
- Shipping temperature:Ice
- Immunogen:HPV Oncoprotein E7, Type 16
- Purification:purified by Protein G chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: HPV 16 (E7)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: