Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Enterococcus species Rabbit Polyclonal Antibody
Anti-Enterococcus species Rabbit Polyclonal Antibody
Catalog # BIRBORB23117-1
Supplier:  Biorbyt
Anti-Enterococcus species Rabbit Polyclonal Antibody
Catalog # BIRBORB23117-1
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Enterococcus sp.
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Storage buffer:
    0,01 M PBS, pH 7,2
  • Storage temperature:
    Cool
  • Concentration:
    4-5 mg/ml (OD280, E(0,1%)=1,4)
  • Shipping temperature:
    Ice
  • Immunogen:
    Enterococcus faecium
  • Purification:
    purified by Protein A chromatography
  • Size:
    1 ml
  • Pk:
    1 mL

Specifications

About this item

Type: Primary
Antigen: Enterococcus species
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: