Anti-C3C Rabbit Polyclonal Antibody
Katalogové číslo BIRBORB21217-1
Dodavatel: Biorbyt
Specifikace
- Antibody type:Primary
- Antigen symbol:C3C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Format:antiserum
- Form:lyophilized
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 ml
- Pk:1 mL
Specifikace
O tomto produktu
Type: Primary
Antigen: C3C
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human