Anti-Secretory Component Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
BIRBORB21195-0.2
: Biorbyt
- Antibody type:Primary
- Antigen symbol:Secretory Component
- Clonality:Monoclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Format:immunoconjugate
- Form:lyophilized
- Storage temperature:Cool
- Concentration:0,4 mg/ml, Peroxidase/IgG protein molar ratio (E/P) approximately 1,7, No foreign proteins added,
- Shipping temperature:Ice
- Immunogen:Highly purified secretory component isolated from human milk.
- Size:200 μg
- Pk:0,2 mg
Type: Primary
Antigen: Secretory Component
Clonality: monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human