Anti-Phospho-(Enol)Pyruvate Carboxylase Rabbit Polyclonal Antibody
Catalog # BIRBORB20968-10
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:EnolPyruvate Carboxylase
- Antigen symbol:Phospho-
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Plant
- Storage buffer:phosphate buffered saline
- Storage temperature:Cool
- Concentration:1 mg/ml, no foreign proteins added,
- Shipping temperature:Ice
- Immunogen:Phospho(enol)pyruvate carboxylase isolated and purified from corn. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Size:10 mg
- Pk:10 mg
Specifications
About this item
Type: Primary
Antigen: Phospho-(Enol)Pyruvate Carboxylase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Plant