Anti-p44/42 MAP Kinase Rabbit Polyclonal Antibody
BIRBORB14625-100
: Biorbyt
- Antibody type:Primary
- Antigen name:p44/42 MAP Kinase
- Antigen symbol:p44/42 MAPK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Storage buffer:PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:The antiserum was produced against synthesized non-phosphopeptide derived from human p44/42 MAP Kinase around the phosphorylation site of tyrosine 204 (T -E-YP-V-A).
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Western Blot: 1:500-1:1000
Type: Primary
Antigen: p44/42 MAP Kinase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat