Anti-ARSH Mouse Polyclonal Antibody
ABNOH00347527-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Arylsulfatase H
- Antigen symbol:ARSH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:347527
- Antigen synonyms:Arylsulfatase H|ARSH_HUMAN.|ASH
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: ARSH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human