Anti-PRPS1L1 Mouse Monoclonal Antibody [clone: 5E10]
Catalog # ABNOH00221823-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Phosphoribosyl Pyrophosphate Synthetase 1-like 1
- Antigen symbol:PRPS1L1
- Clonality:Monoclonal
- Clone:5E10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:221823
- Antigen synonyms:PRPSL|PRS-III|PRPS3|PRPS1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: PRPS1L1
Clonality: Monoclonal
Clone: 5E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat