Anti-ZIC4 Mouse Monoclonal Antibody [clone: 4F3]
ABNOH00084107-M15
: Abnova
- Antibody type:Primary
- Antigen name:Zic Family member 4
- Antigen symbol:ZIC4
- Clonality:Monoclonal
- Clone:4F3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:84107
- Antigen synonyms:Zinc finger protein of the cerebellum 4|ZIC4|Zinc finger protein ZIC 4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: ZIC4
Clonality: Monoclonal
Clone: 4F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human