Anti-PCYOX1 Mouse Monoclonal Antibody [clone: 3D7-D8]
Catalog # ABNOH00051449-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Prenylcysteine oxidase 1
- Antigen symbol:PCYOX1
- Clonality:Monoclonal
- Clone:3D7-D8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:51449
- Antigen synonyms:PCL1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: PCYOX1
Clonality: Monoclonal
Clone: 3D7-D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human