Anti-AGO2 Mouse Monoclonal Antibody [clone: 2E12-1C9]
Catalog # ABNOH00027161-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Argonaute RISC catalytic component 2
- Antigen symbol:AGO2
- Clonality:Monoclonal
- Clone:2E12-1C9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- ChIP:Yes
- Gene ID:27161
- Antigen synonyms:Q10|EIF2C2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: AGO2
Clonality: Monoclonal
Clone: 2E12-1C9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human