Anti-DDX20 Mouse Monoclonal Antibody [clone: 2A12]
Catalog # ABNOH00011218-M04A
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:DEAD (Asp-Glu-Ala-Asp) box polypeptide 20
- Antigen symbol:DDX20
- Clonality:Monoclonal
- Clone:2A12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:11218
- Antigen synonyms:GEMIN3|DP103
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: DDX20
Clonality: Monoclonal
Clone: 2A12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human