Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CLIC3 Mouse Monoclonal Antibody [clone: 2C5]
Anti-CLIC3 Mouse Monoclonal Antibody [clone: 2C5]
Catalog # ABNOH00009022-M04
Supplier:  Abnova
undefined
Anti-CLIC3 Mouse Monoclonal Antibody [clone: 2C5]
Catalog # ABNOH00009022-M04
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Chloride Intracellular Channel 3
  • Antigen symbol:
    CLIC3
  • Clonality:
    Monoclonal
  • Clone:
    2C5
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    9022
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: CLIC3
Clonality: Monoclonal
Clone: 2C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human