Anti-BSND Mouse Monoclonal Antibody [clone: 1E8]
Catalog # ABNOH00007809-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:barttin CLCNK-type chloride channel accessory beta subunit
- Antigen symbol:BSND
- Clonality:Monoclonal
- Clone:1E8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:7809
- Antigen synonyms:DFNB73|BART
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: BSND
Clonality: Monoclonal
Clone: 1E8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human