Order Entry
Czech Republic
ContactUsLinkComponent
Anti-RAN Mouse Polyclonal Antibody
Anti-RAN Mouse Polyclonal Antibody
Catalog # ABNOH00005901-A01
Supplier:  Abnova
undefined
Anti-RAN Mouse Polyclonal Antibody
Catalog # ABNOH00005901-A01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    RAs-related Nuclear protein
  • Antigen symbol:
    RAN
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    5901
  • Antigen synonyms:
    OK/SW-cl.81|GTP-binding nuclear protein Ran|RAN|ARA24|GTPase Ran|Androgen receptor-associated protein 24|Ras-like protein TC4
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µl
  • Pk:
    50 µl

Specifications

About this item

Type: Primary
Antigen: RAN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human