Anti-PSMA4 Mouse Monoclonal Antibody [clone: 2A10-E4]
Catalog # ABNOH00005685-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Proteasome 20S Alpha 4
- Antigen symbol:PSMA4
- Clonality:Monoclonal
- Clone:2A10-E4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5685
- Antigen synonyms:macropain) subunit|proteasome subun|PSMA4|PSC9|HC9|macropain subunit C9|MGC12467|alpha type|multicatalytic endopeptidase complex subunit C9|HsT17706|proteasome (prosome|MGC111191|4|proteasome alpha 4 subunit|MGC24813|proteasome component C9
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: PSMA4
Clonality: Monoclonal
Clone: 2A10-E4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human