Anti-JAG2 Mouse Monoclonal Antibody [clone: 4F10]
Catalog # ABNOH00003714-M03
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Jagged 2
- Antigen symbol:JAG2
- Clonality:Monoclonal
- Clone:4F10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:3714
- Antigen synonyms:mJagged2-1|Serh|HJ2|D12Ggc2e|SER2|sm
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: JAG2
Clonality: Monoclonal
Clone: 4F10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human