Anti-GPD2 Mouse Monoclonal Antibody [clone: 2C11-1F4]
Catalog # ABNOH00002820-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Glycerol-3-phosphate dehydrogenase 2 (mitochondrial)
- Antigen symbol:GPD2
- Clonality:Monoclonal
- Clone:2C11-1F4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:2820
- Antigen synonyms:GDH2|mGPDH|GPDM
- Storage buffer:In 1x PBS, pH 7,4
- Sequence:MAFQKAVKGTLVGGGALATVLGLSQFAHYRRKQMNLAYVKAADCISEPVNREPPSREAQLLTLQNTSEFDILVIGGGATGSGCALDAVTRGLKTALVERDDFSSGTSSRSTKLIHGGVRYLQKAIMKLDIEQYRMVKEALHERANLLEIAPHLSAPLPIMLPVYKWWQLPYYWVGIKLYDLVAGSNCLKSSYVLSKSRALEHFPMLQKDKLVGAIVYYDGQHNDARMNLAIALTAARYGAATANYMEVVSLLKKTDPQTGKVHVSGARCKDVLTGQEFDVRAKCVINATGPFTDSVRKMDDKDAAAICQPSAGVHIVMPGYYSPESMGLLDPATSDGRVIFFLPWQKMTIAGTTDTPTDVTHHPIPSEEDINFILNEV
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Shipping temperature:Ice
- Immunogen:GPD2 (AAH19874, 1 a.a. ~ 378 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against a full length recombinant GPD2.
Recommended Dilutions: Western Blot (1 - 5 µg/ml)
Type: Primary
Antigen: GPD2
Clonality: Monoclonal
Clone: 2C11-1F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human