Anti-CLIC2 Mouse Polyclonal Antibody
ABNOH00001193-B01
: Abnova
- Antibody type:Primary
- Antigen name:Chloride Intracellular Channel 2
- Antigen symbol:CLIC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:1193
- Antigen synonyms:MRXS32|CLIC2b|XAP121
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Type: Primary
Antigen: CLIC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human