Anti-CLIC1 Mouse Monoclonal Antibody [clone: 2D4]
Catalog # ABNOH00001192-M01J
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Chloride Intracellular Channel 1
- Antigen symbol:CLIC1
- Clonality:Monoclonal
- Clone:2D4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:1192
- Antigen synonyms:Homo sapiens chloride intracellular channel 1 (CLIC1)|FLJ94671|CLIC1|Chloride intracellular channel 1 (cDNA|mRNA)
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: CLIC1
Clonality: Monoclonal
Clone: 2D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human