Anti-CLCN2 Mouse Monoclonal Antibody [clone: 3E1]
Catalog # ABNOH00001181-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Chloride Channel 2
- Antigen symbol:CLCN2
- Clonality:Monoclonal
- Clone:3E1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- ChIP:Yes
- Gene ID:1181
- Antigen synonyms:LKPAT|EGMA|EJM6|CLC2|ECA2|CIC-2|ECA3|EGI11|EGI3|EJM8|clC-2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: CLCN2
Clonality: Monoclonal
Clone: 3E1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human