Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CLCA1 Mouse Monoclonal Antibody [clone: 1C4]
Anti-CLCA1 Mouse Monoclonal Antibody [clone: 1C4]
Catalog # ABNOH00001179-M02
Supplier:  Abnova
undefined
Anti-CLCA1 Mouse Monoclonal Antibody [clone: 1C4]
Catalog # ABNOH00001179-M02
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    chloride channel accessory 1
  • Antigen symbol:
    CLCA1
  • Clonality:
    Monoclonal
  • Clone:
    1C4
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    1179
  • Antigen synonyms:
    CaCC-1|CACC1|CLCRG1|GOB5|hCLCA1|hCaCC-1|CACC
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: CLCA1
Clonality: Monoclonal
Clone: 1C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human